Learn More
Ansell™ Trellchem™ Light Protective Suits
Soft, durable material with good resistance to a wide range of industrial chemicals, especially acids and alkalis
Manufacturer:  Ansell™ 478361033
 View more versions of this product
Catalog No. 19-810-109
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Provides excellent protection against hazardous chemicals in liquid, vapor, gaseous, and solid form
  • Strong and flexible polyamide fabric coated with PVC on both sides
  • Large visor made of impact and chemical resistant 2mm PVC material
  • Nitrile/Chloroprene rubber gloves with rubber safety cuffs; attached with the Trellchem Bayonet glove ring system for quick and simple glove exchange
  • Available with attached PVC boots or integrated socks in the suit material
  • Strong and durable gastight zipper closes downwards and is protected by a splash guard
  • Minimum 5-year shelf-life
  • Fully repairable
  • Orange
  • Encapsulating design with hump; breathing apparatus worn inside the suit
  • Ventilation system provides a constant level of overpressure; 2, 30, and 100L/min rates plus zero/off position
  • Large thumbwheel with grooves for a good grip
  • Chemical resistant regulation valve
Certifications
EN 943-1 certified.
Warranty and Services
Two year
Specifications
Specifications
EN 943-1
Large
Gas-tight zipper
Integrated socks in the suit material, impact and chemical-resistant 2mm PVC large visor, Nitrile/Chloroprene Rubber gloves with Rubber safety cuffs, attached with the Trellchem Bayonet glove ring system, ventilation system, large thumbwheel with grooves
Orange
Protection against hazardous chemicals in liquid, vapor, gaseous and solid form
Attached socks
Polyamide Fabric Coated with PVC