Learn More
Ansell™ Trellchem™ Light Protective Suits
Soft, durable material with good resistance to a wide range of industrial chemicals, especially acids and alkalis
Manufacturer:  Ansell™ LG
 View more versions of this product
Catalog No. 19-810-115
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Provides excellent protection against hazardous chemicals in liquid, vapor, gaseous, and solid form
  • Strong and flexible polyamide fabric coated with PVC on both sides
  • Large visor made of impact and chemical resistant 2mm PVC material
  • Nitrile/Chloroprene rubber gloves with rubber safety cuffs; attached with the Trellchem Bayonet glove ring system for quick and simple glove exchange
  • Available with attached PVC boots or integrated socks in the suit material
  • Strong and durable gastight zipper closes downwards and is protected by a splash guard
  • Minimum 5-year shelf-life
  • Fully repairable
  • Orange
  • Encapsulating design with hump; breathing apparatus worn inside the suit
  • Ventilation system provides a constant level of overpressure; 2, 30, and 100L/min rates plus zero/off position
  • Large thumbwheel with grooves for a good grip
  • Chemical resistant regulation valve
Certifications
EN 943-1 certified.
Warranty and Services
Two year
Specifications
Specifications
EN 943-1
Orange
Protection against hazardous chemicals in liquid, vapor, gaseous and solid form
Attached boots
Polyamide Fabric Coated with PVC
Glove ring
Large
Gas-tight zipper
Attached PVC boots, impact and chemical-resistant 2mm PVC large visor, Nitrile/Chloroprene Rubber gloves with Rubber safety cuffs, attached with the Trellchem Bayonet glove ring system, ventilation system, large thumbwheel with grooves