Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell™ Trellchem™ Light Protective Suits

Soft, durable material with good resistance to a wide range of industrial chemicals, especially acids and alkalis

Specifications

Certifications/Compliance EN 943-1
Closure Type Gas-tight zipper
Color Orange
Material Polyamide Fabric Coated with PVC
For Use With (Application) Protection against hazardous chemicals in liquid, vapor, gaseous and solid form
View More Specs

Products 12
Catalog Number Mfr. No. Description Size Price Quantity & Availability  
Catalog Number Mfr. No. Description Size Price Quantity & Availability  
19-810-114 View Documents Ansell™
MD
Attached PVC boots, impact and chemical-resistant 2mm PVC large visor, Nitrile/Chloroprene Rubber gloves with Rubber safety cuffs, attached with the Trellchem Bayonet glove ring system, ventilation system, large thumbwheel with grooves Medium This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-810-115 View Documents Ansell™
LG
Attached PVC boots, impact and chemical-resistant 2mm PVC large visor, Nitrile/Chloroprene Rubber gloves with Rubber safety cuffs, attached with the Trellchem Bayonet glove ring system, ventilation system, large thumbwheel with grooves Large This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-810-116 View Documents Ansell™
XL
Attached PVC boots, impact and chemical-resistant 2mm PVC large visor, Nitrile/Chloroprene Rubber gloves with Rubber safety cuffs, attached with the Trellchem Bayonet glove ring system, ventilation system, large thumbwheel with grooves X-Large This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-810-113 View Documents Ansell™
SM
Attached PVC boots, impact and chemical-resistant 2mm PVC large visor, Nitrile/Chloroprene Rubber gloves with Rubber safety cuffs, attached with the Trellchem Bayonet glove ring system, ventilation system, large thumbwheel with grooves Small This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-810-117 View Documents Ansell™
2XL
Attached PVC boots, impact and chemical-resistant 2mm PVC large visor, Nitrile/Chloroprene Rubber gloves with Rubber safety cuffs, attached with the Trellchem Bayonet glove ring system, ventilation system, large thumbwheel with grooves 2X-Large This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-810-118 View Documents Ansell™
3XL
Attached PVC boots, impact and chemical-resistant 2mm PVC large visor, Nitrile/Chloroprene Rubber gloves with Rubber safety cuffs, attached with the Trellchem Bayonet glove ring system, ventilation system, large thumbwheel with grooves 3X-Large This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-810-107 View Documents Ansell™
478361031
Integrated socks in the suit material, impact and chemical-resistant 2mm PVC large visor, Nitrile/Chloroprene Rubber gloves with Rubber safety cuffs, attached with the Trellchem Bayonet glove ring system, ventilation system, large thumbwheel with grooves Small This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-810-110 View Documents Ansell™
478361034
Integrated socks in the suit material, impact and chemical-resistant 2mm PVC large visor, Nitrile/Chloroprene Rubber gloves with Rubber safety cuffs, attached with the Trellchem Bayonet glove ring system, ventilation system, large thumbwheel with grooves X-Large This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-810-109 View Documents Ansell™
478361033
Integrated socks in the suit material, impact and chemical-resistant 2mm PVC large visor, Nitrile/Chloroprene Rubber gloves with Rubber safety cuffs, attached with the Trellchem Bayonet glove ring system, ventilation system, large thumbwheel with grooves Large This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-810-111 View Documents Ansell™
478361035
Integrated socks in the suit material, impact and chemical-resistant 2mm PVC large visor, Nitrile/Chloroprene Rubber gloves with Rubber safety cuffs, attached with the Trellchem Bayonet glove ring system, ventilation system, large thumbwheel with grooves 2X-Large This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-810-112 View Documents Ansell™
478361041
Integrated socks in the suit material, impact and chemical-resistant 2mm PVC large visor, Nitrile/Chloroprene Rubber gloves with Rubber safety cuffs, attached with the Trellchem Bayonet glove ring system, ventilation system, large thumbwheel with grooves 3X-Large This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-810-108 View Documents Ansell™
478361032
Integrated socks in the suit material, impact and chemical-resistant 2mm PVC large visor, Nitrile/Chloroprene Rubber gloves with Rubber safety cuffs, attached with the Trellchem Bayonet glove ring system, ventilation system, large thumbwheel with grooves Medium This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

  • Provides excellent protection against hazardous chemicals in liquid, vapor, gaseous, and solid form
  • Strong and flexible polyamide fabric coated with PVC on both sides
  • Large visor made of impact and chemical resistant 2mm PVC material
  • Nitrile/Chloroprene rubber gloves with rubber safety cuffs; attached with the Trellchem Bayonet glove ring system for quick and simple glove exchange
  • Available with attached PVC boots or integrated socks in the suit material
  • Strong and durable gastight zipper closes downwards and is protected by a splash guard
  • Minimum 5-year shelf-life
  • Fully repairable
  • Orange

  • Encapsulating design with hump; breathing apparatus worn inside the suit
  • Ventilation system provides a constant level of overpressure; 2, 30, and 100L/min rates plus zero/off position
  • Large thumbwheel with grooves for a good grip
  • Chemical resistant regulation valve

Certifications

EN 943-1 certified.

Warranty and Services

Two year

Specifications

Specifications

EN 943-1
Orange
Protection against hazardous chemicals in liquid, vapor, gaseous and solid form
Gas-tight zipper
Polyamide Fabric Coated with PVC
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit