Learn More
Trellchem™ Light Protective Suits
Soft, durable material with good resistance to a wide range of industrial chemicals, especially acids and alkalis
Manufacturer:  Trelleborg 478361034
 View more versions of this product
Catalog No. 19-810-110
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
EN 943-1
Integrated socks in the suit material, impact and chemical-resistant 2mm PVC large visor, Nitrile/Chloroprene Rubber gloves with Rubber safety cuffs, attached with the Trellchem Bayonet glove ring system, ventilation system, large thumbwheel with grooves
Attached socks
X-Large
Gas-tight zipper
Polyamide Fabric Coated with PVC
Orange
Protection against hazardous chemicals in liquid, vapor, gaseous and solid form