Trellchem™ Light Protective Suits Available on GSA/VA Contract for Federal Government customers only.

Soft, durable material with good resistance to a wide range of industrial chemicals, especially acids and alkalis

Manufacturer: Trelleborg 478361035

 View more versions of this product

Catalog No. 19-810-111

  Check Availability
Add to Cart

    Specifications

    Certifications/Compliance EN 943-1
    Color Orange
    For Use With (Application) Protection against hazardous chemicals in liquid, vapor, gaseous and solid form
    Material Polyamide Fabric Coated with PVC
    Includes Integrated socks in the suit material, impact and chemical-resistant 2mm PVC large visor, Nitrile/Chloroprene Rubber gloves with Rubber safety cuffs, attached with the Trellchem Bayonet glove ring system, ventilation system, large thumbwheel with grooves
    Size 2X-Large
    Closure Type Gas-tight zipper

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit