Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell™ Trellchem™ Light Protective Suits

Catalog Number 19810112
Click to view available options
Description:
Attached PVC boots, impact and chemical-resistant 2mm PVC large visor, Nitrile/Chloroprene Rubber gloves with Rubber safety cuffs, attached with the Trellchem Bayonet glove ring system, ventilation system, large thumbwheel with grooves
Integrated socks in the suit material, impact and chemical-resistant 2mm PVC large visor, Nitrile/Chloroprene Rubber gloves with Rubber safety cuffs, attached with the Trellchem Bayonet glove ring system, ventilation system, large thumbwheel with grooves
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Soft, durable material with good resistance to a wide range of industrial chemicals, especially acids and alkalis

  • Provides excellent protection against hazardous chemicals in liquid, vapor, gaseous, and solid form
  • Strong and flexible polyamide fabric coated with PVC on both sides
  • Large visor made of impact and chemical resistant 2mm PVC material
  • Nitrile/Chloroprene rubber gloves with rubber safety cuffs; attached with the Trellchem Bayonet glove ring system for quick and simple glove exchange
  • Available with attached PVC boots or integrated socks in the suit material
  • Strong and durable gastight zipper closes downwards and is protected by a splash guard
  • Minimum 5-year shelf-life
  • Fully repairable
  • Orange

  • Encapsulating design with hump; breathing apparatus worn inside the suit
  • Ventilation system provides a constant level of overpressure; 2, 30, and 100L/min rates plus zero/off position
  • Large thumbwheel with grooves for a good grip
  • Chemical resistant regulation valve

Certifications

EN 943-1 certified.

Warranty and Services

Two year

Certifications/Compliance EN 943-1
Color Orange
For Use With (Application) Protection against hazardous chemicals in liquid, vapor, gaseous and solid form
Material PVC-coated Polyamide
Size 3X-Large
Ankle Style Attached socks
Closure Type Gas-tight zipper
Description Integrated socks in the suit material, impact and chemical-resistant 2mm PVC large visor, Nitrile/Chloroprene Rubber gloves with Rubber safety cuffs, attached with the Trellchem Bayonet glove ring system, ventilation system, large thumbwheel with grooves
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit