Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ansell™ Trellchem™ Light Protective Suits
Click to view available options
Description:
Attached PVC boots, impact and chemical-resistant 2mm PVC large visor, Nitrile/Chloroprene Rubber gloves with Rubber safety cuffs, attached with the Trellchem Bayonet glove ring system, ventilation system, large thumbwheel with grooves
Integrated socks in the suit material, impact and chemical-resistant 2mm PVC large visor, Nitrile/Chloroprene Rubber gloves with Rubber safety cuffs, attached with the Trellchem Bayonet glove ring system, ventilation system, large thumbwheel with grooves
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Description
- Provides excellent protection against hazardous chemicals in liquid, vapor, gaseous, and solid form
- Strong and flexible polyamide fabric coated with PVC on both sides
- Large visor made of impact and chemical resistant 2mm PVC material
- Nitrile/Chloroprene rubber gloves with rubber safety cuffs; attached with the Trellchem Bayonet glove ring system for quick and simple glove exchange
- Available with attached PVC boots or integrated socks in the suit material
- Strong and durable gastight zipper closes downwards and is protected by a splash guard
- Minimum 5-year shelf-life
- Fully repairable
- Orange
- Encapsulating design with hump; breathing apparatus worn inside the suit
- Ventilation system provides a constant level of overpressure; 2, 30, and 100L/min rates plus zero/off position
- Large thumbwheel with grooves for a good grip
- Chemical resistant regulation valve
Certifications
EN 943-1 certified.
Warranty and Services
Two year
Specifications
Certifications/Compliance | EN 943-1 |
Color | Orange |
For Use With (Application) | Protection against hazardous chemicals in liquid, vapor, gaseous and solid form |
Material | PVC-coated Polyamide |
Size | 2X-Large |
Ankle Style | Attached boots |
Closure Type | Gas-tight zipper |
Cuff Style | Locking glove ring |
Description | Attached PVC boots, impact and chemical-resistant 2mm PVC large visor, Nitrile/Chloroprene Rubber gloves with Rubber safety cuffs, attached with the Trellchem Bayonet glove ring system, ventilation system, large thumbwheel with grooves |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction