Learn More
Ansell™ Trellchem™ Pressure Test Kits and Accessories
Contains everything required to make a quick and simple check of the gastight integrity of Level A/Type 1 gastight chemical protective suits. Options include kits for testing Encapsulating suits (TE), Non-encapsulating suits (T) and Combination kits for b
Manufacturer:  Ansell™ 487091000
 View more versions of this product
Catalog No. 19-810-242
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Alert:
All suits shoulld be inspected after every use or at least once every year including gastightness test, checking the suit material and components for damage and lubricating the zipper.
Description
Description
Trelltest allows for testing according to the European test method EN 464 (stipulated in EN 943) as well as ASTM F 1052 (stipulated n NFPA 1991).
  • Contents include:
    • 1 pc adapter (kit TE) / 1 pc face seal plate (kit T), with connection nipple and valve
    • 1 pc pressure gauge with PVC hose and quick release coupling
    • 1 pc digital stop watch/timer
    • 3 pcs (kit TE) / 2 pcs (kit T) rubber blind plugs
    • 2 pcs cuff clips (used with old ring system)
    • All packed in a black plastic box
  • Accessories include: Unidaptor Universal Test Kit Adaptor and Suit Inflation System
Specifications
Specifications
Suit
Uniadapter