Learn More
Ansell™ Trellchem™ Pressure Test Kits and Accessories
Click to view available options
Product Type:
Combination Encapsulated/Nonencapsulated Test Kit
Encapsulated Test Kit
Nonencapsulated Test Kit
Description
Trelltest allows for testing according to the European test method EN 464 (stipulated in EN 943) as well as ASTM F 1052 (stipulated n NFPA 1991).
- Contents include:
- 1 pc adapter (kit TE) / 1 pc face seal plate (kit T), with connection nipple and valve
- 1 pc pressure gauge with PVC hose and quick release coupling
- 1 pc digital stop watch/timer
- 3 pcs (kit TE) / 2 pcs (kit T) rubber blind plugs
- 2 pcs cuff clips (used with old ring system)
- All packed in a black plastic box
- Accessories include: Unidaptor Universal Test Kit Adaptor and Suit Inflation System
Specifications
For Use With (Equipment) | Suit |
Product Type | Encapsulated Test Kit |