Trellchem™ Pressure Test Kits
Manufacturer: Trelleborg 487090078
 View more versions of this product
Catalog No. 19-810-236
  Check Availability
Add to Cart
Alert
All suits shoulld be inspected after every use or at least once every year including gastightness test, checking the suit material and components for damage and lubricating the zipper.
    Specifications
    Size Combination Encapsulated/Nonencapsulated Test Kit
    For Use With (Equipment) Suit
  • Description & Specifications
Related Products
Product Content Feedback