Learn More
Trellchem™ Repair Supplies
To supplement Trellchem Complete Repair Kits for protective suits
Manufacturer:  Trelleborg 487080071
 View more versions of this product
Catalog No. 19-810-223
  Check Availability
Add to cart
Alert:
All suits shouldl be inspected after every use or at least once every year including gastightness test, checking the suit material and components for damage and lubricating the zipper.
Specifications Description & Specifications
Specifications
Repair patches
8/Pk.
To supplement Trellchem Complete Repair Kits
For repair of light protective suits