Learn More
Trellchem™ Repair Supplies
To supplement Trellchem Complete Repair Kits for protective suits
Manufacturer:  Trelleborg 001721203
 View more versions of this product
Catalog No. 19-810-230
  Check Availability
Add to cart
Alert:
All suits shouldl be inspected after every use or at least once every year including gastightness test, checking the suit material and components for damage and lubricating the zipper.
Specifications Description & Specifications
Specifications
Trellchem Solvent
250mL
To supplement Trellchem Complete Repair Kits
For repair of Trellchem protective suits