Learn More
Ansell™ Trellchem™ Super and Super Freeflow Suits
Description
Offer maximum strength and durability with retained softness and flexibility.
- Provides excellent protection against hazardous chemicals in liquid, vapor, gaseous and solid form
- Combination of butyl rubber with Viton™ offers outstanding resistance to chemical attack from a wide range of chemicals
- Trellchem Viton/butyl rubber gloves with rubber safety cuffs; attached with the Trellchem Bayonet glove ring system for quick and simple glove exchange
- Both available with attached socks of the suit material; Super Freeflow also available with black nitrile rubber safety boots
- Strong and durable gastight zipper closes downwards and is protected by a splash guard
- Minimum 10-year shelf-life
- Fully repairable
- Yellow
Super Suits
- Feature large, VP1 visor made of impact and chemical resistant 2mm PVC material
- Encapsulating design with hump; breathing apparatus worn inside the suit
- Ventilation system provides a constant level of overpressure; 2, 30, and 100L/min rates plus zero/off position
- Large thumbwheel with grooves for a good grip
- Chemical resistant regulation valve
- Large, VP1 visor made of impact and chemical resistant 2mm PVC material
- Feature smaller CV visor made of impact and chemical resistant 2mm PVC material
- Encapsulating design without hump; connects to an external air source via swiveling passthrough on the back of the suit
- No SCBA use; the air inside the suit is the breathing air
- Air distributes through two diffusers/silencers at the top of the hood to prevent fogging of the visor
- Working Pressure: 3-8 bars
- Warning whistle sounds below 3 bars (220 l/min).
- Airflow: 220 l/min (minimum); 475 l/min (maximum)
•Breakthrough Time (Minutes)
Greater than 480: Acetone; Acetonitrile; Anhydrous ammonia; 1, 3 Butadiene; Carbon disulfide 95%; Chlorine; Dimethyl formamide; Diethyl amine; Ethylene oxide; Heptane; Hexane; Hydrogen chloride; Methanol; Methyl chloride; Mustard gas; Nitrobenzene; Sodium hydroxide 40%; Sulphuric acid 98%; VX
Greater than 1400: Lewisite; Sarine; Soman; Tabun
Dichloromethane: 58
Diethyl amine: 53
Ethyl acetate: 252
Tetrachloroethylene: 143
Tetrahydrofuran: 28
Toluene: 74
Certifications
EN 943-1 certified.
Warranty and Services
Two year
Specifications
Certifications/Compliance | EN 943-1 |
Color | Yellow |
Material | Elastomer-coated Woven Fabric |
Size | Large |
Ankle Style | Attached socks |
Closure Type | Gas-tight zipper |
Description | Attached socks of the suit material, 2mm PVC large VP1 visor, Trellchem Viton™/Butyl Rubber gloves with Rubber safety cuffs, attached with the Trellchem Bayonet glove ring system, ventilation system, large thumbwheel with grooves; Encapsulating design without hump |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.