Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell™ Trellchem™ Super and Super Freeflow Suits

Super material based on real woven fabric and coated with elastomers

Specifications

Certifications/Compliance EN 943-1
Closure Type Gas-tight zipper
Color Yellow
Material Real Woven Fabric and Coated with Elastomers
View More Specs

Products 18
Catalog Number Mfr. No. Description Size Price Quantity & Availability  
Catalog Number Mfr. No. Description Size Price Quantity & Availability  
19-810-101 View Documents Ansell™
490360041BOOT
Black Nitrile Rubber safety boots, 2mm CV small VP1 visor, Trellchem Viton™/Butyl Rubber gloves with Rubber safety cuffs, attached with the Trellchem Bayonet glove ring system, ventilation system, large thumbwheel with grooves; Encapsulating design without hump Small This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-810-102 View Documents Ansell™
490360042BOOT
Black Nitrile Rubber safety boots, 2mm CV small VP1 visor, Trellchem Viton™/Butyl Rubber gloves with Rubber safety cuffs, attached with the Trellchem Bayonet glove ring system, ventilation system, large thumbwheel with grooves; Encapsulating design without hump Medium This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-810-103 View Documents Ansell™
490360043BOOT
Black Nitrile Rubber safety boots, 2mm CV small VP1 visor, Trellchem Viton™/Butyl Rubber gloves with Rubber safety cuffs, attached with the Trellchem Bayonet glove ring system, ventilation system, large thumbwheel with grooves; Encapsulating design without hump Large This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-810-083 View Documents Ansell™
490360031
Attached socks of the suit material, 2mm PVC large VP1 visor, Trellchem Viton™/Butyl Rubber gloves with Rubber safety cuffs, attached with the Trellchem Bayonet glove ring system, ventilation system, large thumbwheel with grooves; Encapsulating design without hump Small This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-810-104 View Documents Ansell™
490360044BOOT
Black Nitrile Rubber safety boots, 2mm CV small VP1 visor, Trellchem Viton™/Butyl Rubber gloves with Rubber safety cuffs, attached with the Trellchem Bayonet glove ring system, ventilation system, large thumbwheel with grooves; Encapsulating design without hump X-Large This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-810-105 View Documents Ansell™
490360045BOOT
Black Nitrile Rubber safety boots, 2mm CV small VP1 visor, Trellchem Viton™/Butyl Rubber gloves with Rubber safety cuffs, attached with the Trellchem Bayonet glove ring system, ventilation system, large thumbwheel with grooves; Encapsulating design without hump 2X-Large This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-810-084 View Documents Ansell™
490360032
Attached socks of the suit material, 2mm PVC large VP1 visor, Trellchem Viton™/Butyl Rubber gloves with Rubber safety cuffs, attached with the Trellchem Bayonet glove ring system, ventilation system, large thumbwheel with grooves; Encapsulating design without hump Medium This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-810-106 View Documents Ansell™
490360049BOOT
Black Nitrile Rubber safety boots, 2mm CV small VP1 visor, Trellchem Viton™/Butyl Rubber gloves with Rubber safety cuffs, attached with the Trellchem Bayonet glove ring system, ventilation system, large thumbwheel with grooves; Encapsulating design without hump 3X-Large This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-810-086 View Documents Ansell™
490360034
Attached socks of the suit material, 2mm PVC large VP1 visor, Trellchem Viton™/Butyl Rubber gloves with Rubber safety cuffs, attached with the Trellchem Bayonet glove ring system, ventilation system, large thumbwheel with grooves; Encapsulating design without hump X-Large This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-810-087 View Documents Ansell™
490360035
Attached socks of the suit material, 2mm PVC large VP1 visor, Trellchem Viton™/Butyl Rubber gloves with Rubber safety cuffs, attached with the Trellchem Bayonet glove ring system, ventilation system, large thumbwheel with grooves; Encapsulating design without hump 2X-Large This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-810-085 View Documents Ansell™
490360033
Attached socks of the suit material, 2mm PVC large VP1 visor, Trellchem Viton™/Butyl Rubber gloves with Rubber safety cuffs, attached with the Trellchem Bayonet glove ring system, ventilation system, large thumbwheel with grooves; Encapsulating design without hump Large This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-810-096 View Documents Ansell™
490360042
Attached socks of the suit material, 2mm PVC small VP1 visor, Trellchem Viton™/Butyl Rubber gloves with Rubber safety cuffs, attached with the Trellchem Bayonet glove ring system, ventilation system, large thumbwheel with grooves; Encapsulating design without hump Medium This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-810-095 View Documents Ansell™
490360044
Attached socks of the suit material, 2mm PVC small VP1 visor, Trellchem Viton™/Butyl Rubber gloves with Rubber safety cuffs, attached with the Trellchem Bayonet glove ring system, ventilation system, large thumbwheel with grooves; Encapsulating design without hump X-Large This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-810-097 View Documents Ansell™
490360043
Attached socks of the suit material, 2mm PVC small VP1 visor, Trellchem Viton™/Butyl Rubber gloves with Rubber safety cuffs, attached with the Trellchem Bayonet glove ring system, ventilation system, large thumbwheel with grooves; Encapsulating design without hump Large This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-810-088 View Documents Ansell™
490360041
Attached socks of the suit material, 2mm PVC large VP1 visor, Trellchem Viton™/Butyl Rubber gloves with Rubber safety cuffs, attached with the Trellchem Bayonet glove ring system, ventilation system, large thumbwheel with grooves; Encapsulating design without hump 3X-Large This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-810-098 View Documents Ansell™
490360047
Attached socks of the suit material, 2mm PVC small VP1 visor, Trellchem Viton™/Butyl Rubber gloves with Rubber safety cuffs, attached with the Trellchem Bayonet glove ring system, ventilation system, large thumbwheel with grooves; Encapsulating design without hump X-Large This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-810-099 View Documents Ansell™
490360045
Attached socks of the suit material, 2mm PVC small VP1 visor, Trellchem Viton™/Butyl Rubber gloves with Rubber safety cuffs, attached with the Trellchem Bayonet glove ring system, ventilation system, large thumbwheel with grooves; Encapsulating design without hump 2X-Large This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-810-100 View Documents Ansell™
490360049
Attached socks of the suit material, 2mm PVC small VP1 visor, Trellchem Viton™/Butyl Rubber gloves with Rubber safety cuffs, attached with the Trellchem Bayonet glove ring system, ventilation system, large thumbwheel with grooves; Encapsulating design without hump 3X-Large This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Offer maximum strength and durability with retained softness and flexibility.

  • Provides excellent protection against hazardous chemicals in liquid, vapor, gaseous and solid form
  • Combination of butyl rubber with Viton™ offers outstanding resistance to chemical attack from a wide range of chemicals
  • Trellchem Viton/butyl rubber gloves with rubber safety cuffs; attached with the Trellchem Bayonet glove ring system for quick and simple glove exchange
  • Both available with attached socks of the suit material; Super Freeflow also available with black nitrile rubber safety boots
  • Strong and durable gastight zipper closes downwards and is protected by a splash guard
  • Minimum 10-year shelf-life
  • Fully repairable
  • Yellow

Super Suits

  • Feature large, VP1 visor made of impact and chemical resistant 2mm PVC material
  • Encapsulating design with hump; breathing apparatus worn inside the suit
  • Ventilation system provides a constant level of overpressure; 2, 30, and 100L/min rates plus zero/off position
  • Large thumbwheel with grooves for a good grip
  • Chemical resistant regulation valve
  • Large, VP1 visor made of impact and chemical resistant 2mm PVC material
Super Freeflow Suits
  • Feature smaller CV visor made of impact and chemical resistant 2mm PVC material
  • Encapsulating design without hump; connects to an external air source via swiveling passthrough on the back of the suit
  • No SCBA use; the air inside the suit is the breathing air
  • Air distributes through two diffusers/silencers at the top of the hood to prevent fogging of the visor
  • Working Pressure: 3-8 bars
  • Warning whistle sounds below 3 bars (220 l/min).
  • Airflow: 220 l/min (minimum); 475 l/min (maximum)

Breakthrough Time (Minutes)
Greater than 480: Acetone; Acetonitrile; Anhydrous ammonia; 1, 3 Butadiene; Carbon disulfide 95%; Chlorine; Dimethyl formamide; Diethyl amine; Ethylene oxide; Heptane; Hexane; Hydrogen chloride; Methanol; Methyl chloride; Mustard gas; Nitrobenzene; Sodium hydroxide 40%; Sulphuric acid 98%; VX
Greater than 1400: Lewisite; Sarine; Soman; Tabun
Dichloromethane: 58
Diethyl amine: 53
Ethyl acetate: 252
Tetrachloroethylene: 143
Tetrahydrofuran: 28
Toluene: 74

Certifications

EN 943-1 certified.

Warranty and Services

Two year

Specifications

Specifications

EN 943-1
Gas-tight zipper
Yellow
Real Woven Fabric and Coated with Elastomers
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit