Trellchem™ Super and Super Freeflow Suits Available on GSA/VA Contract for Federal Government customers only.
Super material based on real woven fabric and coated with elastomers
Manufacturer: Trelleborg 490360031
 View more versions of this product
Catalog No. 19-810-083
  Check Availability
Add to Cart
    Specifications
    Certifications/Compliance EN 943-1
    Material Real Woven Fabric and Coated with Elastomers
    Includes Attached socks of the suit material, 2mm PVC large VP1 visor, Trellchem Viton™/Butyl Rubber gloves with Rubber safety cuffs, attached with the Trellchem Bayonet glove ring system, ventilation system, large thumbwheel with grooves
    Color Yellow
    Size Small
    Disposable No
    Closure Type Gas-tight zipper
    Construction Encapsulating design without hump
  • Description & Specifications
Related Products
Product Content Feedback