Learn More
Trellchem™ Super and Super Freeflow Suits Available on GSA/VA Contract for Federal Government customers only.
Super material based on real woven fabric and coated with elastomers
Manufacturer:  Trelleborg 490360043BOOT
 View more versions of this product
Catalog No. 19-810-103
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
EN 943-1
Encapsulating design without hump
Black Nitrile Rubber safety boots, 2mm CV small VP1 visor, Trellchem Viton™/Butyl Rubber gloves with Rubber safety cuffs, attached with the Trellchem Bayonet glove ring system, ventilation system, large thumbwheel with grooves
Large
Gas-tight zipper
Real Woven Fabric and Coated with Elastomers
Yellow