Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell™ Trellchem™ Super-T Level B Suits

Super material based on real woven fabric coated with elastomers; maximum strength and durability with retained softness and flexibility

Manufacturer:  Ansell™ 490220014

 View more versions of this product

Catalog No. 19-810-092

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Includes: Minihood

Description

Description

  • Provides excellent protection against hazardous chemicals in liquid, vapor, gaseous and solid form
  • Combination of butyl rubber with Viton™ offers outstanding resistance to chemical attack from a wide range of chemicals
  • Trellchem Viton/butyl rubber gloves with rubber safety cuffs; attached with the Trellchem Bayonet glove ring system for quick and simple glove exchange
  • Attached socks of the suit material
  • Rubber face seal is anatomically designed for optimum safety and comfort
  • Strong and durable gastight zipper closes downwards and is protected by a splash guard
  • Minimum 10-year shelf-life
  • Fully repairable

  • Nonencapsulating design with hump; breathing apparatus worn inside the suit
  • Ventilation system provides a constant level of overpressure; 2, 30, and 100L/min rates plus zero/off position
  • Large thumbwheel with grooves for a good grip
  • Chemical resistant regulation valve

·Breakthrough Time (Minutes)
Greater than 480: Acetone; Acetonitrile; Anhydrous ammonia; 1, 3 Butadiene; Carbon disulfide 95%; Chlorine; Dimethyl formamide; Diethyl amine; Ethylene oxide; Heptane; Hexane; Hydrogen chloride; Methanol; Methyl chloride; Mustard gas; Nitrobenzene; Sodium hydroxide 40%; Sulphuric acid 98%; VX
Greater than 1400: Lewisite; Sarine; Soman; Tabun
Dichloromethane: 58
Diethyl amine: 53
Ethyl acetate: 252
Tetrachloroethylene: 143
Tetrahydrofuran: 28
Toluene: 74

Note: To fulfill EN 943-1 standard, TC hood or minihood is required.

Certifications

EN 943-1 certified.

Warranty and Services

Two year

Specifications

Specifications

EN 943-1
X-Large
Non-encapsulating
No
Yellow
Provides excellent protection against hazardous chemicals in liquid, vapor, gaseous and solid form
Gas-tight zipper with splash guard
Viton™, Butyl Rubber
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit