Learn More
Trellchem™ Super-T Level B Suits Available on GSA/VA Contract for Federal Government customers only.
Super material based on real woven fabric coated with elastomers; maximum strength and durability with retained softness and flexibility
Manufacturer:  Trelleborg 490220011
 View more versions of this product
Catalog No. 19-810-089
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
EN 943-1
Viton™, Butyl Rubber
Small
Provides excellent protection against hazardous chemicals in liquid, vapor, gaseous and solid form
Gas-tight zipper with splash guard
Yellow
No
Non-encapsulating