Learn More
Ansell™ Trellchem™ Super-T Level B Suits
Description
Includes
Minihood
- Provides excellent protection against hazardous chemicals in liquid, vapor, gaseous and solid form
- Combination of butyl rubber with Viton™ offers outstanding resistance to chemical attack from a wide range of chemicals
- Trellchem Viton/butyl rubber gloves with rubber safety cuffs; attached with the Trellchem Bayonet glove ring system for quick and simple glove exchange
- Attached socks of the suit material
- Rubber face seal is anatomically designed for optimum safety and comfort
- Strong and durable gastight zipper closes downwards and is protected by a splash guard
- Minimum 10-year shelf-life
- Fully repairable
- Nonencapsulating design with hump; breathing apparatus worn inside the suit
- Ventilation system provides a constant level of overpressure; 2, 30, and 100L/min rates plus zero/off position
- Large thumbwheel with grooves for a good grip
- Chemical resistant regulation valve
•Breakthrough Time (Minutes)
Greater than 480: Acetone; Acetonitrile; Anhydrous ammonia; 1, 3 Butadiene; Carbon disulfide 95%; Chlorine; Dimethyl formamide; Diethyl amine; Ethylene oxide; Heptane; Hexane; Hydrogen chloride; Methanol; Methyl chloride; Mustard gas; Nitrobenzene; Sodium hydroxide 40%; Sulphuric acid 98%; VX
Greater than 1400: Lewisite; Sarine; Soman; Tabun
Dichloromethane: 58
Diethyl amine: 53
Ethyl acetate: 252
Tetrachloroethylene: 143
Tetrahydrofuran: 28
Toluene: 74
Note: To fulfill EN 943-1 standard, TC hood or minihood is required.
Certifications
EN 943-1 certified.
Warranty and Services
Two year
Specifications
Certifications/Compliance | EN 943-1 |
Color | Yellow |
For Use With (Application) | Provides excellent protection against hazardous chemicals in liquid, vapor, gaseous and solid form |
Material | Viton™, Butyl Rubber |
Size | 3X-Large |
Closure Type | Gas-tight zipper with splash guard |
Disposable | No |
Style | Non-encapsulating |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.