Learn More
Ansell™ Trellchem™ VPS-Flash NPFA 1991-2005 Certified Suits
Certified with chemical flash fire and liquefied gas options
Manufacturer:  Ansell™ 476520091
 View more versions of this product
Catalog No. 19-810-071
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Combination of elastomers and plastics with a woven fabric and Nomex for chemical flash fire protection. Maximum protection against hazardous chemicals in liquid, vapor, gaseous and solid form, including warfare agents.
  • Encapsulating design with hump; breathing apparatus worn inside the suit
  • Strong, flexible material offers an outstanding chemical barrier—more than 8 hours protection against a wide range of chemicals
  • Chloroprene rubber outer layer provides excellent flame as well as abrasion resistance; no overcover required
  • Seam stiched with aramide thread for superior strength and durability; taped with a rubber strip on the outside and a barrier laminate strip welded to the inside for a continuous barrier layer across the seam
  • Gloves consist of three layers: An inner 4H SilverShield™ barrier glove; an outer chloroprene rubber glove; and a removable outer Kevlar™ glove; Trellchem Bayonet glove ring system offers quick and simple glove exchange (Kevlar glove providing mechanical protection is to be worn over the glove assembly)
  • Strong and durable gastight zipper closes downwards and is protected by a splash guard
  • Integrated bootie in the primary suit material (user must wear separate outer safety boots to cover the bootie)
  • Large visor made of impact and chemical resistant, 2mm PVC material
  • Fully repairable
  • Orange
·Breakthrough Time (Minutes)
Greater than 480: Acetone; Acetonitrile; Anhydrous ammonia; 1, 3 Butadiene; Carbon disulfide 95%; Chlorine; Dichloromethane; Diethyl amine; Dimethyl formamide; Ethyl acetate; Ethylene oxide; Heptane; Hexane; Hydrogen chloride; Methanol; Methyl chloride; Nitrobenzene; Sodium hydroxide 40%; Sulphuric acid 98%; Tetrachloroethylene; Tetrahydrofuran; Toluene
Greater than 1400: Lewisite; Mustard gas; Sarine; Soman; Tabun; VX
Certifications
NFPA 1991-2005 certified, including optional chemical flash fire and liquefied gas protection requirements.
Warranty and Services
Two year
Specifications
Specifications
NFPA 1991-2005
Small
Attached socks
Orange
Gas-tight zipper
Integrated bootie in the primary suit material, 2mm PVC large visor, inner 4H™ SilverShield™/outer Chloroprene Rubber/removable outer Dupont™ Kevlar™ aramid fiber glove, Trellchem Bayonet glove ring system; Combination of Elastomers and Plastics with woven Fabric and Dupont™ Nomex™ aramid fibers for chemical flash fire protection, encapsulating design with hump