Trellchem™ VPS-Flash NPFA 1991-2005 Certified Suits Available on GSA/VA Contract for Federal Government customers only.
Certified with chemical flash fire and liquefied gas options
Manufacturer: Trelleborg 476520093
 View more versions of this product
Catalog No. 19-810-073
  Check Availability
Add to Cart
    Specifications
    Certifications/Compliance NFPA 1991-2005
    Material PVC
    Includes Integrated bootie in the primary suit material, 2mm PVC large visor, inner 4H™ SilverShield™/outer Chloroprene Rubber/removable outer Kevlar™ glove, Trellchem Bayonet glove ring system
    Color Orange
    Size Large
    Closure Type Gas-tight zipper
    Construction Combination of Elastomers and Plastics with woven Fabric and Nomex™ for chemical flash fire protection, encapsulating design with hump
  • Description & Specifications
Related Products
Product Content Feedback