Learn More
Trelleborg™ ONEGlove™ Hazmat Gloves
Protect against chemical/biological agents, industrial solvents and chemicals, cuts and punctures, and flash fires
Manufacturer:  Trelleborg 072250501
 View more versions of this product
Catalog No. 19-810-307
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
OEM Specific
Protects against cuts, chemical/biological agents, chemicals, flash fires
Small
NFPA 1991, NFPA 1994