Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell™ Trelleborg™ Silicone Oversock

Increases ease of sliding protective boots on and off

Manufacturer:  Ansell™ 072210100

Catalog No. 19-810-200

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

  • Size: One size fits all
  • Material: Silicone
  • Color: Red

For use with all Trellchem Level A and Level B suits

Specifications

Specifications

Silicone
Universal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit