Learn More
Ansell™ Trelleborg™ Trellchem™ Trellcover Cryogenic and Frostbite Protective Gloves
Protective apparel minimizes frostbite risk
Manufacturer:  Ansell™ 474590001
Catalog No. 19-810-183
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Includes: One pair lined outer gloves
Description
Description
Coated polyamide fabric provides good flexibility in low temperatures.
Does not provide chemical protection. To be worn over a chemical protective suit when working with condensed gases, such as liquid chlorine and ammonia.
Warranty and Services
Two year