Learn More
Ansell™ Trelltrain™ Level A Training Suits
Level A training suit of strong and flexible polyamide fabric, coated with PVC on both sides
Manufacturer:  Ansell™ ZXL
 View more versions of this product
Catalog No. 19-810-160
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Highly abrasion resistant
  • Nitrile/Chloroprene rubber gloves with rubber safety cuffs; attached with the Trellchem Bayonet glove ring system for quick and simple glove exchange
  • Large visor made of impact- and chemical-resistant 2mm PVC material
  • Material and visor are fully repairable
  • Watertight zipper protected by splash guard
  • Integrated sock/bootie in the suit material
  • Encapsulating design with hump; breathing apparatus worn inside the suit
  • Ventilation system provides a constant level of overpressure; 2, 30, and 100L/min rates plus zero/off position
  • Large thumbwheel with grooves for a good grip
  • Chemical resistant regulation valve
Warranty and Services
Two year
Specifications
Specifications
Green
Zipper
Integrated sock/bootie in the suit material, 2mm PVC large visor, Nitrile/Chloroprene Rubber glove with Rubber safety cuffs, Trellchem Bayonet glove ring system, ventilation system, large thumbwheel with grooves; Encapsulating design with hump
X-Large
Attached socks
Polyamide Fabric Coated with PVC