Learn More
Ansell™ Trelltrain Training Suit
A suit intended for training only but designed in the same way as your real gastight intervention suit, to be able to train as realistically as possible.
Manufacturer:  Ansell™ TRELLTRAIN
Catalog No. 19-820-720
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Trelltrain comes in three different designs to fit all user preferences: Encapsulating design type CV, Encapsulating design type VP1 (larger visor) and Non-encapsulating design type T
  • Non-certified product designed for realistic training
  • Strong, soft and durable fabric coated with PVC on both sides
  • Saves the operational suits until they are really needed
  • Same design as the operational Trellchem suit
  • Welded seams and durable watertight PVC coated zipper