Learn More
TREM-1 (CD354) Mouse anti-Human, Brilliant Violet 711, Clone: 6B1, BD Optibuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 743742
Description
The 6B1monoclonal antibody specifically binds to TREM-1 (Triggering Receptor Expressed on Myeloid Cells 1 or Triggering Receptor Expressed on Monocytes 1) which is also known as CD354. CD354 is a type I transmembrane glycoprotein that belongs to the immunoglobulin superfamily. CD354 is highly expressed by neutrophils and monocytes. It is also reportedly expressed at lower levels by a variety of cells including subsets of T and B cells, NK cells and dendritic cells. Its expression is upregulated on leucocytes in response to microbial lipopolysaccharide, eg, during microbial infections or sepsis. CD354 activation triggers the release of proinflammatory cytokines and chemokines. A soluble form of CD354 has been described that may play a protective role in some infectious disease states.
The antibody was conjugated to BD Horizon™ BV711 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 711nm. BD Horizon BV711 can be excited by the violet laser and detected in a filter used to detect Cy™5.5 / Alexa Fluor™ 700-like dyes (eg, 712/20nm filter). Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy5.5 detectors. However, the spillover can be corrected through compensation as with any other dye combination. BV711 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 711 nm. This dye offers a very bright choice for the violet laser. Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy5.5 detectors. BV711 will also have moderate spillover into the BD Horizon Brilliant™ Violet 786 detector.
Specifications
TREM-1 (CD354) | |
6B1 | |
Brilliant Violet 711 | |
TREM1, Triggering receptor expressed on myeloid cells 1 | |
Human Trem-1 Recombinant Protein | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
IgG1 κ |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.