Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TREM1 Mouse anti-Human, Alexa Fluor 532, Clone: MM0583-9T3, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP211977AF532
Description
TREM1 Monoclonal specifically detects TREM1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
TREM1 | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
TREM1 | |
Human recombinant TREM1 (extracellular domain) | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
MM0583-9T3 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
CD354 antigen, TREM-1Triggering receptor expressed on monocytes 1, triggering receptor expressed on myeloid cells 1, triggering-receptor TREM1 | |
Mouse | |
Protein G purified | |
Signal Transduction | |
54210.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title