Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TREM-2 ectodomain Rat anti-Human, Mouse, Clone: 5F4, MilliporeSigma™

Rat Monoclonal Antibody

$145.80 - $373.00

Specifications

Antigen TREM-2 ectodomain
Clone 5F4
Concentration 1.0 mg/mL
Applications Western Blot
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABN2321100
View Documents
MilliporeSigma
MABN2321100UG
100 μg
Each for $373.00
Add to cart
 
MABN232125
View Documents
MilliporeSigma
MABN232125UG
25 μg
Each for $145.80
Add to cart
 
Description

Description

TREM-2 ectodomain Monoclonal antibody specifically detects TREM-2 ectodomain in Human, Mouse samples. It is validated for Western Blotting.
Specifications

Specifications

TREM-2 ectodomain
1.0 mg/mL
Unconjugated
Rat
Human, Mouse
His-tagged recombinant extracellular domain of murine TREM-2.
Monoclonal
Protein G purified
5F4
Western Blot
Purified
RUO
Purified rat monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
IgG2aκ
Primary
Stable for 1 year at +2°C to +8°C from date of receipt.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit