Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TREM-2 Rat anti-Mouse, Unconjugated, Clone: 14D1, MilliporeSigma™

Rat Monoclonal Antibody

$153.50 - $386.00

Specifications

Antigen TREM-2
Applications Immunohistochemistry (Paraffin), Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rat
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABN23201MI
View Documents
MilliporeSigma
MABN2320100UG
100 μg
Each for $386.00
Add to cart
 
MABN23202MI
View Documents
MilliporeSigma
MABN232025UG
25 μg
Each for $153.50
Add to cart
 
Description

Description

TREM-2 Monoclonal antibody specifically detects TREM-2 in Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blotting.
Specifications

Specifications

TREM-2
Unconjugated
Rat
Triggering receptor expressed on myeloid cells 2;Triggering receptor expressed on monocytes 2
A linear peptide corresponding to 20 amino acids from the C-terminal region of murine Triggering receptor expressed on myeloid cells 2 (TREM-2).
Primary
Stable for 1 year at 2 to 8°C from date of receipt.
Immunohistochemistry (Paraffin), Western Blot
Purified
Mouse
Trem2, Trem2a, Trem2c
Monoclonal
Protein G Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit