Learn More
TREM2b Rat anti-Mouse, DyLight 405, Clone: RM0139-5J46, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP122515V
Catalog No. NBP122515V
Add to cart
Description
Description
TREM2b Monoclonal antibody specifically detects TREM2b in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
TREM2b
RM0139-5J46
Western Blot
Purified
TREM2
Mouse recombinant protein of the extracellular domain of TREM-2b
Protein G purified
Store at 4C in the dark.
Monoclonal
This was produced from a hybridoma (mouse myeloma) fused with spleen cells from a rat immunized with mouse recombinant protein of the EC-domain of TREM-2b.
Western Blot
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
Trem2a, Trem2b, Trem2c, TREM-2triggering receptor expressed on myeloid cells 2a, Triggering receptor expressed on monocytes 2, triggering receptor expressed on myeloid cells 2
Rat
IgG2
0.1 ml
Primary
54209
Mouse