Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ TREM2 Recombinant Human Protein, His Tag Non-distribution product as customer accommodation.

Expressed from DNA sequence encoding extracellular domain of human TREM2 (Met 1-Ser 174) fused to a polyhistidine tag at the C-terminus.

Manufacturer:  Thermo Scientific™ 11084H08H50

Catalog No. 11-084-H08H50


Add to cart

Description

Description

Expressed from DNA sequence encoding extracellular domain of human TREM2 (Met 1-Ser 174) fused to a polyhistidine tag at the C-terminus.

  • Lyophilized in PBS, pH7.4, 5% mannitol, 5% trehalose, 0.02% Tween™-80
Specifications

Specifications

Analysis of protein structure, used as immunogen, protein standard, or in cell biology research applications
54209
≥85% by SDS-PAGE
Human
<1 EU/μg
TREM2
Other proteins
TREM2 Recombinant Human Protein
Lyophilized
18.9kd
50μg
Truncated, Recombinant, Mutant
Triggering Receptor Expressed On Myeloid Cells 2
Assay Standard
Mammalian
His Tag
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only. Not for use in diagnostic procedures.