Learn More
Abnova™ TREML1 (Human) ELISA Kit
Sandwich enzyme-linked immunosorbent assay
Manufacturer:  Abnova™ KA5170
Catalog No. 89-335-206
Add to cart
Description
Description
TREML1 (Human) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for the quantitative measurement of human TREML1 in cell culture supernates, cell lysates, serum and plasma (heparin, EDTA).
Specifications
Specifications
Human
Quantitative
RUO
1 Kit
triggering receptor expressed on myeloid cells-like 1
340205
Store at 4°C for 6 months, at -20°C for 12 months. Avoid multiple freeze-thaw cycles.
Human