Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TREX1 Mouse anti-Human, Alexa Fluor™ 594, Clone: 41M5F2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP222394AF594

Catalog No. NP222394A94


Add to cart

Description

Description

TREX1 Monoclonal antibody specifically detects TREX1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

TREX1
41M5F2
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Conjugated, Purified
TREX1
Within the range of amino acids 211-350 of human TREX1 were used as immunogen for the antibody.
Protein G purified
DNA Repair, Editing and Processing Endonucleases
Primary
11277.0
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
3′-5′ exonuclease TREX1, AGS1, Aicardi-Goutieres syndrome 1, CRV, DKFZp434J0310, DNase III, DRN3, EC 3.1.11.2, HERNS, three prime repair exonuclease 1,3′ repair exonuclease 1
Mouse
IgG2b Kappa
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit