Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRF-1 Mouse anti-Human, Janelia Fluor 646, Clone: 40000, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP141217JF646

Catalog No. NBP14217646


Add to cart

Description

Description

TRF-1 Monoclonal antibody specifically detects TRF-1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

TRF-1
40000
Immunocytochemistry/Immunofluorescence
Purified
TERF1
The exact sequence is proprietary.
Protein G purified
Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Core ESC Like Genes, Stem Cell Markers
Primary
7013
Human
Immunocytochemistry, Immunofluorescence
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
NIMA-interacting protein 2, PIN2hTRF1-AS, Telomeric protein Pin2/TRF1, telomeric repeat binding factor (NIMA-interacting) 1, telomeric repeat binding factor 1, telomeric repeat binding protein 1, telomeric repeat-binding factor 1, TERF1, TRBF1, TRF1FLJ41416, TRFt-TRF1, TTAGGG repeat-binding factor 1
Mouse
IgG1
0.1mL
Store at 4°C in the dark.
Monoclonal
This antibody will recognize human TRF1.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit