Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRF-2, Mouse anti-Human, Alexa Fluor 594, Clone: 3H6B4, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP226298AF594

Catalog No. NB005368

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

TRF-2 Monoclonal antibody specifically detects TRF-2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

TRF-2
3H6B4
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Conjugated
TERF2
Partial recombinant human TRF2 (between residues 300-500) expressed in E. coli [Uniprot# Q15554]
Ammonium sulfate precipitation
RUO
Store at 4°C in the dark.
Monoclonal
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Alexa Fluor 594
50mM Sodium Borate with 0.05% Sodium Azide
Telomeric DNA-binding protein, telomeric repeat binding factor 2, Telomeric repeat binding protein 2, telomeric repeat-binding factor 2, TERF 2, TERF2, TRBF 2, TRBF2, TRF 2, TRF2, TTAGGG repeat binding factor 2, TTAGGG repeat-binding factor 2
Mouse
IgG1
0.1 mL
Cancer, Cellular Markers, Chromatin Research, Transcription Factors and Regulators, Tumor Suppressors
Primary
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit