Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRF Goat anti-Mouse, Polyclonal , Abnova™

Goat polyclonal antibody raised against mouse Trf.

Manufacturer:  Abnova Corporation PAB29692

Catalog No. 89-935-051

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Specifications

Specifications

TRF
Unconjugated
Affinity Purified
Trf
Native purified mouse Trf.
Antigen Affinity Purification
RUO
Primary
22041
ELISA
In PBS, pH 7.2 (0.09% sodium azide)
AI266983, Cd176, HP, MGC102653, Tfn, hpx
Goat
IgG
1mg
Store at 4°C. Aliquot to avoid repeated freezing and thawing.
Polyclonal
Murine
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit