Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRF1 Rat anti-Mouse, Unconjugated, Clone: 572C, MilliporeSigma™
Rat Monoclonal Antibody
Supplier: MilliporeSigma MABT1321100UG
Description
TRF1 Monoclonal antibody specifically detects TRF1 in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blotting.Specifications
TRF1 | |
Monoclonal | |
Unconjugated | |
Telomeric repeat-binding factor 1;TTAGGG repeat-binding factor | |
Rat | |
∼51 kDa observed; 48.22 kDa calculated. Uncharacterized bands may be observed in some lysate(s). | |
100 μg | |
Primary | |
Mouse | |
Purified |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
572C | |
NP_033378.1 | |
Tefrf1, Trf1 | |
His-tagged full-length recombinant mouse Telomeric repeat-binding factor 1 (TRF1). | |
Protein G Purified | |
Epigenetics and Nuclear Function | |
Clone 572C is a rat monoclonal antibody that specifically detects murine Telomeric repeat-binding factor 1 (TRF1). | |
Stable for 1 year at 2 to 8°C from date of receipt. | |
IgG1 κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction