Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRF1 Rat anti-Mouse, Unconjugated, Clone: 572C, MilliporeSigma™

Rat Monoclonal Antibody

$153.50 - $386.00

Specifications

Antigen TRF1
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rat
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABT13211MI
View Documents
MilliporeSigma
MABT1321100UG
100 μg
Each for $386.00
Add to cart
 
MABT13212MI
View Documents
MilliporeSigma
MABT132125UG
25 μg
Each for $153.50
Add to cart
 
Description

Description

TRF1 Monoclonal antibody specifically detects TRF1 in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blotting.
Specifications

Specifications

TRF1
Unconjugated
Rat
Telomeric repeat-binding factor 1;TTAGGG repeat-binding factor
His-tagged full-length recombinant mouse Telomeric repeat-binding factor 1 (TRF1).
Primary
Stable for 1 year at 2 to 8°C from date of receipt.
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
Purified
Mouse
Tefrf1, Trf1
Monoclonal
Protein G Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit