Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRF2 Rabbit anti-Human, Mouse, Rat, Polyclonal, FabGennix

Rabbit Polyclonal Antibody
Supplier: Fabgennix Inc TRF2201AP

Description
TRF2 Polyclonal antibody specifically detects TRF2 in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western BlotSpecifications
TRF2 | |
Polyclonal | |
Unconjugated | |
TERF2 | |
Telomeric DNA-binding protein; telomeric repeat binding factor 2; telomeric repeat binding protein 2; telomeric repeat-binding factor 2; TERF2; TRBF2; TRF2; TTAGGG repeat-binding factor 2 | |
Synthetic peptide taken within amino acid region 80-130 on human Telomeric repeat-binding factor 2 protein. | |
200 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
0.5-1.5 mg/mL | |
proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8 | |
O35144, Q15554 | |
Rabbit | |
Affinity chromatography | |
RUO | |
21750, 361403, 7014 | |
-20°C | |
Liquid |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title