Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TRH Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557331
Description
Immunogen sequence: HKRQHPGRRE DEASWSVDVT QHKRQHPGRR SPWLAYAVPK RQHPGRRLAD PKAQRSWEE Highest antigen sequence identity to the following orthologs: Mouse - 51%, Rat - 50%.
This gene encodes a member of the thyrotropin-releasing hormone family. Cleavage of the encoded proprotein releases mature thyrotropin-releasing hormone, which is a tripeptide hypothalamic regulatory hormone. The human proprotein contains six thyrotropin-releasing hormone tripeptides. Thyrotropin-releasing hormone is involved in the regulation and release of thyroid-stimulating hormone, as well as prolactin. Deficiency of this hormone has been associated with hypothalamic hypothyroidism.
Specifications
TRH | |
Polyclonal | |
Unconjugated | |
TRH | |
Prothyroliberin; Pro-thyrotropin-releasing hormone; protirelin; Pro-TRH; THR; Thyroliberin; thyrotropin releasing hormone; thyrotropin-releasing factor; Thyrotropin-releasing hormone; TRF; Trh; TRH01; TSH-releasing factor | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
7200 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.50 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P20396 | |
TRH | |
Recombinant protein corresponding to Human TRH. Recombinant protein control fragment (Product #RP-96454). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction