Learn More
Invitrogen™ TRH Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA597006
Description
Positive Samples: SH-SY5Y, Mouse brain Immunogen sequence: QPEAAQQEAV TAAEHPGLDD FLRQVERLLF LRENIQRLQG DQGEHSASQI FQSDWLSKRQ HPGKREEEEE EGVEEEEEEE GGAVGPHKRQ HPGRREDEAS WSVDVTQHKR QHPGRRSPWL AYAVPKRQHP GRRLADPKAQ RSWEEEEEEE EREEDLMPEK RQHPGKRALG GPCGPQGAYG QAGLLLGLLD DLSRSQGAEE KRQHPG.
This gene encodes a member of the thyrotropin-releasing hormone family. Cleavage of the encoded proprotein releases mature thyrotropin-releasing hormone, which is a tripeptide hypothalamic regulatory hormone. The human proprotein contains six thyrotropin-releasing hormone tripeptides. Thyrotropin-releasing hormone is involved in the regulation and release of thyroid-stimulating hormone, as well as prolactin. Deficiency of this hormone has been associated with hypothalamic hypothyroidism.
Specifications
TRH | |
Polyclonal | |
Unconjugated | |
TRH | |
Prothyroliberin; Pro-thyrotropin-releasing hormone; protirelin; Pro-TRH; THR; Thyroliberin; thyrotropin releasing hormone; thyrotropin-releasing factor; Thyrotropin-releasing hormone; TRF; Trh; TRH01; TSH-releasing factor | |
Rabbit | |
Affinity Chromatography | |
RUO | |
22044, 7200 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
2.09 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
P20396, Q62361 | |
TRH | |
A synthetic peptide corresponding to a sequence within amino acids 25-230 of human TRH (NP_0090481). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.