Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRH Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549509

Catalog No. PA549509

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Functions as a regulator of the biosynthesis of TSH in the anterior pituitary gland and as a neurotransmitter/ neuromodulator in the central and peripheral nervous systems. May promote hair shaft elongation, prolonge the hair cycle growth phase (anagen) and antagonized its termination by TGFB2. May also increase proliferation and inhibited apoptosis of hair matrix keratinocytes.
Specifications

Specifications

TRH
Polyclonal
PBS with 0.09% sodium azide
P20396
Pro-thyrotropin-releasing hormone, Pro-TRH, Prothyroliberin, Thyrotropin-releasing hormone, TRH, Protirelin, TSH-releasing factor, Thyroliberin, Thyrotropin-releasing factor, TRF, TRH
Rabbit
IgG
200 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
7200
Western Blot
Unconjugated
TRH
Liquid
TRH
KLH conjugated synthetic peptide between 157-191 amino acids from human TRH.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit