Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tri(1-naphthyl)phosphine, +96%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
3411-48-1
C30H21P
412.472
MFCD00046400
DMEUUKUNSVFYAA-UHFFFAOYSA-N
tri 1-naphthyl phosphine,tri-1-naphthylphosphine,tri naphthalen-1-yl phosphine,tris 1-naphthyl phosphine,tris naphthalen-1-yl phosphane,trinaphthyl phosphine,tri-1-naphtylphosphine,tri-1-napthylphosphine,acmc-1cp9e,tri naphthalen-1-yl phosphane
271443
trinaphthalen-1-ylphosphane
C1=CC=C2C(=C1)C=CC=C2P(C3=CC=CC4=CC=CC=C43)C5=CC=CC6=CC=CC=C65
3411-48-1
C30H21P
412.472
MFCD00046400
DMEUUKUNSVFYAA-UHFFFAOYSA-N
tri 1-naphthyl phosphine,tri-1-naphthylphosphine,tri naphthalen-1-yl phosphine,tris 1-naphthyl phosphine,tris naphthalen-1-yl phosphane,trinaphthyl phosphine,tri-1-naphtylphosphine,tri-1-napthylphosphine,acmc-1cp9e,tri naphthalen-1-yl phosphane
271443
trinaphthalen-1-ylphosphane
C1=CC=C2C(=C1)C=CC=C2P(C3=CC=CC4=CC=CC=C43)C5=CC=CC6=CC=CC=C65
Infrared Spectrum Authentic
Quantity 1g
Color Yellow
Packaging Glass bottle
Melting Point 265.0°C to 268.0°C
Assay Percent Range 96+%
Formula Weight 412.47
Physical Form Powder
Percent Purity ≥96%
Chemical Name or Material Tri(1-naphthyl)phosphine

GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.