Alfa Aesar™ Tri(ethylene glycol) bis(2-ethylhexanoate), 90+%

Manufacturer: Alfa Aesar™ H54406AE

Quantity 100mL

 View more versions of this product

Catalog No. AAH54406AE

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 94-28-0
    Molecular Formula C22H42O6
    Formula Weight 402.57
    MDL Number MFCD00072285
    Chemical Name or Material Tri(ethylene glycol) bis(2-ethylhexanoate)

    Specifications

    Formula Weight 402.57
    TSCA TSCA
    MDL Number MFCD00072285
    CAS 94-28-0
    Boiling Point 344°C
    Melting Point −50°C
    RTECS Number MO7725000
    Quantity 100mL
    Molecular Formula C22H42O6
    Chemical Name or Material Tri(ethylene glycol) bis(2-ethylhexanoate)
    Assay ≥90%

    Description

    Applications
    It is widely used in water based adhesive, painting and coating industry. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and dyestuffs.

    Solubility
    Insoluble in water.

    Notes
    Incompatible with strong oxidizing agents. Store at room temperature.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit