Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tri-Methyl-Histone H3 (Lys36) Rabbit anti-Human, Mouse, Invitrogen™

Rabbit Recombinant Monoclonal Antibody

Manufacturer:  Invitrogen MA524687

Catalog No. PIMA524687


Add to cart

Description

Description

This antibody reacts to Histone H3 trimethylated at Lysine 36 (K36me3). No cross reactivity with monomethylated Lysine 36 (K36me1) or dimethylated Lysine 36 (K36me2), or other methylations in histone H3. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using Recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

Histone H3 is one of the four core proteins of the nucleosome, and it is involved in transcription regulation, DNA repair, DNA replication and chromosomal stability. The N-terminal tail of Histone H3 undergoes many post-translational modifications, including phosphorylation, acetylation, multiple methylation, ubiquitination, and ADP-ribosylation to achieve its diverse functions. HistoneH3 is acetylated and deacetylated on N-terminal lysine residues. Acetylation removes the positive charge on the histone, decreasing the interaction with the negatively charged phosphate groups of DNA, and resulting in a more relaxed structure associated with greater levels of gene transcription. Acetylation of histone H3 at lysine 9 (H3K9Ac) is one of the most well-known epigenetic markers enriched in the promoter region of activated genes.
Specifications

Specifications

Tri-Methyl-Histone H3 (Lys36)
Recombinant Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH 7.2-7.4
P68431, P68433
H3 histone family, member A; H3a; H3K9ac; HIST1H3A; Histone 1; histone 1, H3a; Histone H3.1; Histone H3/a; Histone H3/b; Histone H3/c; Histone H3/d; Histone H3/f; Histone H3/h; Histone H3/i; Histone H3/j; Histone H3/k; Histone H3/l
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
360198, 8350
ChIP assay, ELISA, Immunocytochemistry, Immunofluorescence, Peptide Array, Western Blot
RM155
Unconjugated
HIST1H3A
Liquid
HIST1H3A
Tri-methyl-peptide corresponding to tri-methyl-Histone H3 (Lys36).
Protein A
RUO
Antibody
Monoclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only