Learn More
Tri-Methyl-Histone H3 (Lys9) Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA540069
Catalog No. PA540069
Add to cart
Description
Description
Histones are the main constituents of the protein part of chromosomes of eukaryotic cells. They are rich in the amino acids arginine and lysine and have been greatly conserved during evolution. Histones pack the DNA into tight masses of chromatin. Two core histones of each class H2A, H2B, H3 and H4 assemble and are wrapped by 146 base pairs of DNA to form one octameric nucleosome. Histone tails undergo numerous post-translational modifications, which either directly or indirectly alter chromatin structure to facilitate transcriptional activation or repression or other nuclear processes. In addition to the genetic code, combinations of the different histone modifications reveal the so-called ""histone code"". Histone methylation and demethylation is dynamically regulated by respectively histone methyl transferases and histone demethylases. Trimethylation of histone H3K9 is associated with heterochromatin formation and gene silencing.
Specifications
Specifications
Tri-Methyl-Histone H3 (Lys9)
Polyclonal
whole serum with 0.05% sodium azide
P68431, P68433
H3K9me3, Histone H3, Histone 1, H3a, Histone H3.1, histone H3/a, HIST1H3A
Rabbit
IgG
RUO
Antibody
Polyclonal
Human, Mouse
ELISA, Immunocytochemistry, Immunofluorescence, Peptide Array, Western Blot
Unconjugated
HIST1H3A
Liquid
HIST1H3A
KLH-conjugated synthetic peptide corresponding to human histone H3 containing the trimethylated lysine 9 (H3K9me3).
100 μL
-20°C or -80°C if preferred
Primary
360198, 8350