Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tri-n-butyl(3-methyl-2-butenyl)tin, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
Tri-n-butyl(3-methyl-2-butenyl)tin is used in prenylation of ethyl propiolate. It is also involved in chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Tri-n-butyl(3-methyl-2-butenyl)tin is used in prenylation of ethyl propiolate. It is also involved in chemical research.
Solubility
Immiscible with water.
Notes
Store in a cool place. Incompatible with strong oxidizing agents.
Tri-n-butyl(3-methyl-2-butenyl)tin is used in prenylation of ethyl propiolate. It is also involved in chemical research.
Solubility
Immiscible with water.
Notes
Store in a cool place. Incompatible with strong oxidizing agents.
Specifications
Density | 1.042 |
Boiling Point | 283°C |
Flash Point | >110°C (230°F) |
Refractive Index | 1.488 |
Quantity | 1 g |
UN Number | UN2788 |
Beilstein | 3588742 |
Solubility Information | Immiscible with water. |
Formula Weight | 359.19 |
Percent Purity | 95% |
Show More |
Safety and Handling
Hazard Category | H301-H312-H315-H319-H372 |
Hazard Statement | GHS H Statement H301-H372-H312-H315-H319 Toxic if swallowed. Causes damage to organs through prolonged or repeated exposure. Harmful in contact with skin. Causes skin irritation. Causes serious eye irritation. |
Precautionary Statement | P260-P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c |
DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S. |
TSCA | No |
Recommended Storage | Keep cold |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction