Alfa Aesar™ Tri-n-butyl citrate, 99%

Manufacturer: Alfa Aesar™ A1026618

Quantity 50g

 View more versions of this product

Catalog No. AAA1026618

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 77-94-1
    Molecular Formula C18H32O7
    Formula Weight 360.45
    MDL Number MFCD00027217
    Beilstein 1806072
    Merck Index 14,1564
    Synonym Citric acid tri-n-butyl ester
    Chemical Name or Material Tri-n-butyl citrate

    Specifications

    Formula Weight 360.45
    Flash Point 157°C (315°F)
    Boiling Point 325°C
    TSCA TSCA
    Odor Weak
    MDL Number MFCD00027217
    Beilstein 1806072
    Merck Index 14,1564
    Synonym Citric acid tri-n-butyl ester
    RTECS Number TZ8608000
    Melting Point −20°C
    Quantity 50g
    Molecular Formula C18H32O7
    CAS 77-94-1
    Chemical Name or Material Tri-n-butyl citrate
    Assay 99%

    Description

    Applications
    Tri-n-butyl citrate is used in polishes, inks, nitrocellulose lacquers, as an anti-foam agent and as a plasticizer. It is also used as paint additives, coating additives and processing aids. In addition to this, it is added to the zein film to enhance its mechanical properties for industrial processing.

    Solubility
    Immiscible with water.

    Notes
    Incompatible with strong oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit