Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Tri-n-butyl citrate, 99%

Manufacturer:  Alfa Aesar™ A1026630

Quantity 250g

 View more versions of this product

Catalog No. AAA1026630


  Check Availability
Add to cart

Specifications

Description

Applications
Tri-n-butyl citrate is used in polishes, inks, nitrocellulose lacquers, as an anti-foam agent and as a plasticizer. It is also used as paint additives, coating additives and processing aids. In addition to this, it is added to the zein film to enhance its mechanical properties for industrial processing.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
Description & Specifications

Chemical Identifiers

77-94-1
360.45
1806072
Citric acid tri-n-butyl ester
C18H32O7
MFCD00027217
14,1564
Tri-n-butyl citrate

Specifications

157°C (315°F)
−20°C
77-94-1
C18H32O7
MFCD00027217
14,1564
Tri-n-butyl citrate
325°C
250g
Weak
360.45
1806072
Citric acid tri-n-butyl ester
99%

Safety and Handling

SDS & Safety Documents

RTECSNumber : TZ8608000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit